Anti-RAD50 Mouse Monoclonal Antibody [clone: 10G5]

Supplier: Prosci

47-128
10071-720EA 615.28 USD
10071-720
Anti-RAD50 Mouse Monoclonal Antibody [clone: 10G5]
Antibodies
Protein G-purified

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RAD50
Clonality: Monoclonal
Clone: 10G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1/kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR