Anti-FOXO1 Mouse Monoclonal Antibody

Supplier: Prosci

47-121
10070-964EA 615.28 USD
10070-964
Anti-FOXO1 Mouse Monoclonal Antibody
Antibodies
Protein G-purified

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FOXO1
Clonality: Monoclonal
Clone: 1G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1/kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR