Anti-ESR2 Mouse Monoclonal Antibody [clone: 1C3]

47-120
10070-962EA 813.33 USD
10070-962
Anti-ESR2 Mouse Monoclonal Antibody [clone: 1C3]
Antibodies
Protein G-purified

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ESR2
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1/kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR