Anti-E2F1 Mouse Monoclonal Antibody [clone: D01]

Supplier: Prosci

47-117
10070-942EA 420.98 USD
10070-942
Anti-E2F1 Mouse Monoclonal Antibody [clone: D01]
Antibodies
Protein a-purified

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: E2F1
Clonality: Monoclonal
Clone: D01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR