Anti-CENPF Mouse Monoclonal Antibody [clone: 2B6]

Supplier: Prosci

47-114
10070-952EA 615.28 USD
10070-952
Anti-CENPF Mouse Monoclonal Antibody [clone: 2B6]
Antibodies
Protein G-purified

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CENPF
Clonality: Monoclonal
Clone: 2B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1/kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR