Anti-BRCA2 Mouse Monoclonal Antibody [clone: 1D12]

Supplier: Prosci

47-112
10070-948EA 615.28 USD
10070-948
Anti-BRCA2 Mouse Monoclonal Antibody [clone: 1D12]
Antibodies
Protein G-purified

PBS containing 0.02% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BRCA2
Clonality: Monoclonal
Clone: 1D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1/kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR