Anti-NBN Rabbit Polyclonal Antibody

Supplier: Prosci
47-108
10071-742EA 776.86 USD
10071-742
Anti-NBN Rabbit Polyclonal Antibody
Antibodies
1 mg/ml BSA, 0.05% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NBN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR