Anti-TSPAN8 Goat Polyclonal Antibody

Supplier: Prosci
46-998
10117-438EA 701.94 USD
10117-438
Anti-TSPAN8 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 24 kDa band observed in human colon lysates (calculated MW of 26.0 kDa according to NP_004607.1). Recommended concentration: 0.01-0.03 ?g/mL.

Tetraspanin 8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TSPAN8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR