Anti-IFT57 Goat Polyclonal Antibody

Supplier: Prosci
46-988
10117-420EA 701.94 USD
10117-420
Anti-IFT57 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 49 kDa band observed in human and mouse brain lysates (calculated MW of 49.1 kDa according to human NP_060480.1 and of 48.8 kDa according to mouse NP_082956.2). Recommended concentration: 1-3 ?g/mL.

IFT57 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IFT57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR