Anti-DLX5 Goat Polyclonal Antibody

Supplier: Prosci
46-983
10117-410EA 701.94 USD
10117-410
Anti-DLX5 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 30 kDa band observed in human bone marrow lysates (calculated MW of 31.5 kDa according to NP_005212.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Lee et al, J Biol Chem. 2003 Sep 5;278 (36) :34387-94; PMID: 12815054). Recommended concentration: 1-3 ?g/mL.

DLX5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DLX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR