Anti-HOXB4 Goat Polyclonal Antibody

Supplier: Prosci
46-972
10117-388EA 552.17 USD
10117-388
Anti-HOXB4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Preliminary experiments in lysates of K562 gave no specific signal but low background (at antibody concentration up to 1 ?g/mL).

homeobox B4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HOXB4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR