Anti-AUTS2 Goat Polyclonal Antibody

Supplier: Prosci
46-970
10117-384EA 625.23 USD
10117-384
Anti-AUTS2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Approximately 60 kDa band in human brain (cerebral cortex and frontal cortex) lysates after 0.3 ?g/mL antibody staining.1 and of 136 kDa according to NP_001120703.1. The 60 kDa band was successfully blocked by incubation with the immunizing peptide.

AUTS2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AUTS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR