Anti-Cpt1a Goat Polyclonal Antibody

Supplier: Prosci
46-959
10117-364EA 552.17 USD
10117-364
Anti-Cpt1a Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Preliminary experiments gave next to a 90 kDa band additional bands at approximately 37 kDa and 25 kDa in mouse and rat liver lysates after 0.3 ?g/mL antibody staining.3 kDa according to NP_038523.2. All detected bands were successfully blocked by incubation with the immunizing peptide (and BLAST results with the immunizing peptide sequence did not identify any other proteins to explain the additional bands).

CPT1A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Cpt1a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR