Anti-TRPM8 Goat Polyclonal Antibody

Supplier: Prosci
46-940
10117-326EA 552.17 USD
10117-326
Anti-TRPM8 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Approximately 28 kDa band in lysates of cell line NIH3T3 after 1 ?g/mL antibody staining. The 28 kDa band was successfully blocked by incubation with the immunizing peptide.

TRPM8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TRPM8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR