Anti-MARCH10 Goat Polyclonal Antibody

Supplier: Prosci
46-938
10117-322EA 701.94 USD
10117-322
Anti-MARCH10 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8000. Approximately 90 kDa band observed in rat testis lysates (calculated MW of 90.5 kDa according to human NP_689811.2 and of 87.9 kDa according to mouse NP_766156.2). Recommended concentration: 0.3-1 µg/mL. an additional band of unknown identity was also consistently observed at 37 kDa . This band was successfully blocked by incubation with the immunizing peptide.

MARCH10 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MARCH10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR