Anti-ZEB1 Goat Polyclonal Antibody

Supplier: Prosci
46-932
10117-310EA 701.76 USD
10117-310
Anti-ZEB1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 100 kDa band observed in human heart and mouse heart lysates (calculated MW of 122 kDa according to human NP_110378.3 and mouse NP_035676.1). Recommended concentration: 0.05-0.

ZEB1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ZEB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR