Anti-ARNTL Goat Polyclonal Antibody

Supplier: Prosci
46-924
10117-294EA 701.94 USD
10117-294
Anti-ARNTL Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 70+75 kDa bands observed in human cerebellum lysates (calculated MW of 68.7 kDa according to NP_001169.3 and 64.1 kDa according to NP_001025444.1).

ARNTL antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ARNTL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR