Anti-RSPH1 Goat Polyclonal Antibody

Supplier: Prosci
46-916
10117-280EA 701.94 USD
10117-280
Anti-RSPH1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 37 kDa band observed in mouse testis lysates (calculated MW of 35.1 kDa according to human NP_543136.1 and 34.2 kDa according to mouse NP_079566.1). Recommended concentration: 0.

RSpH 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RSPH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR