Anti-TRPV3 Goat Polyclonal Antibody

46-915
10117-278EA 689.45 USD
10117-278
Anti-TRPV3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 26 kDa band in human skin lysates after 0.3 ?g/mL antibody staining. In transfected HEK293 transiently expressing full-length human TRPV3 (myc and DYKDDDDK tagged), a band of approximately. 90 kDa was observed. No bands were observed in mock-transfected HEK293 and the same band was observed using anti-DYKDDDDK tag antibody. Recommended concentration, 0.5-1 ?g/mL.

TRPV3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TRPV3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR