Anti-TRIB1 Goat Polyclonal Antibody

Supplier: Prosci
46-912
10117-274EA 701.94 USD
10117-274
Anti-TRIB1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 35 kDa band in human lymph node and Peripheral Blood Mononucleocyte lysates after 0.3 ?g/mL antibody staining.

Tribbles Homolog 1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TRIB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR