Anti-TMPRSS4 Goat Polyclonal Antibody

Supplier: Prosci
46-909
10117-268EA 701.94 USD
10117-268
Anti-TMPRSS4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 48 kDa band observed in human bone marrow lysates (calculated MW of 48.2 kDa according to NP_063947.1 and of 47.6 kDa according to NP_001077416.1). Recommended concentration: 1-3

TMPRSS4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TMPRSS4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR