Anti-RAB23 Goat Polyclonal Antibody

Supplier: Prosci
46-889
10113-156EA 701.94 USD
10113-156
Anti-RAB23 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 30 kDa band observed in mouse eye lysates (calculated MW of 26.7 kDa according to human NP_057361.3 and 26.8 kDa according to mouse NP_033025.2).

RAB23 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RAB23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR