Anti-Cma2 Goat Polyclonal Antibody

Supplier: Prosci
46-865
10113-112EA 552.17 USD
10113-112
Anti-Cma2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 49 kDa band in human skeletal muscle lysates after 0.02 ?g/mL antibody staining.

Chymase 2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Cma2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR