Anti-GPR125 Goat Polyclonal Antibody

Supplier: Prosci
46-851
10113-084EA 701.76 USD
10113-084
Anti-GPR125 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:8,000. Western Blot: Approximately 140 kDa band observed in human ovary lysates (calculated MW of 146 kDa according to NP_660333.2). Recommended concentration: 0.5-1.5 ?g/mL.

GPR125 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR125
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR