Anti-GDF2 Goat Polyclonal Antibody

Supplier: Prosci
46-848
10113-080EA 552.17 USD
10113-080
Anti-GDF2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:4,000. Western Blot: Approximately 27 kDa band in human bone marrow, liver, and cerebellum lysates after 1 ?g/mL antibody staining. Please note that currently we canno

GDF2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GDF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR