Anti-DLG1 Goat Polyclonal Antibody

Supplier: Prosci
46-831
10113-048EA 701.94 USD
10113-048
Anti-DLG1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 150 kDa band observed in lysates of cell line KELLY (calculated MW of 103 kDa according to NP_004078.2). The observed molecular weight corresponds to earlier findings with dif

DLG1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DLG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR