Anti-ABCA6 Goat Polyclonal Antibody

Supplier: Prosci
46-790
10117-218EA 552.17 USD
10117-218
Anti-ABCA6 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 50 kDa band in human brain (cerebellum, cerebral cortex, frontal cortex) lysates after 0.5 ?g/mL antibody staining. Please note that current

ABCA6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ABCA6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR