Anti-ABCA2 Goat Polyclonal Antibody

Supplier: Prosci
46-787
10117-214EA 701.94 USD
10117-214
Anti-ABCA2 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2,000. Western Blot: Approximately 35 kDa band in human brain (amygdala, frontal cortex, hippocampus and Substantia Nigra) lysates after 0.3 ?g/mL antibody staining. Please no

ABCA2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ABCA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR