Anti-ANXA1 Goat Polyclonal Antibody

Supplier: Prosci
46-785
10117-210EA 701.94 USD
10117-210
Anti-ANXA1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 35 kDa band observed in human lysates of cell lines A431 and K562 (calculated MW of 38.7 kDa according to NP_000691.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Paweletz et al, tumor Res. 2000 Nov15;60 (22) :6293-7. PMID: 11103786). Recommended concentration: 0.5-1.5 ?g/mL.

Annexin A1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ANXA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR