Anti-ALOX15 Goat Polyclonal Antibody

Supplier: Prosci
46-769
10117-182EA 701.94 USD
10117-182
Anti-ALOX15 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 80 kDa band observed in nuclear HeLa lysate (calculated MW of 74.8 kDa according to NP_001131.3). Recommended concentration: 0.2-0.5 ?g/mL.

ALOX15 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ALOX15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR