Anti-MEIS1 Goat Polyclonal Antibody

Supplier: Prosci
46-767
10117-178EA 701.94 USD
10117-178
Anti-MEIS1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 40 kDa band observed in human cerebral cortex lysates (calculated MW of 43.0 kDa according to NP_002389.1). Recommended concentration: 0.1-0.3 ?g/mL.

MEIS1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MEIS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR