Anti-U2AF1L4 Goat Polyclonal Antibody

Supplier: Prosci
46-759
10117-162EA 701.94 USD
10117-162
Anti-U2AF1L4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 20Da band observed in human cerebellum and lung lysates (calculated MW of 21.3 kDa according to NP_001035515.1). Recommended concentration: 0.1-0.3 ?g/mL.

U2AF1L4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: U2AF1L4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR