Anti-PCSK6 Goat Polyclonal Antibody

Supplier: Prosci
46-729
10117-106EA 701.94 USD
10117-106
Anti-PCSK6 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 40-45 kDa band in human cerebellum lysates after 0.2 ?g/mL antibody staining. Immunohistochemistry: In paraffin embedded human kkeletal muscle shows striated staining in alternating fibres. Recommended concentration, 5-10 ?g/mL.

PCSK6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PCSK6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR