Anti-STRA6 Goat Polyclonal Antibody

Supplier: Prosci
46-715
10117-084EA 701.94 USD
10117-084
Anti-STRA6 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 85 kDa band observed in rat brain lysates (calculated MW of 73.8 kDa according to NP_001025095.1). Recommended concentration: 0.3-1 ?g/mL.

STRA6 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Stra6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR