Anti-RPGRIP1L Goat Polyclonal Antibody

Supplier: Prosci
46-709
10117-074EA 701.94 USD
10117-074
Anti-RPGRIP1L Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 150 kDa band observed in human cerebellum lysates (calculated MW of 151 kDa according to NP_056087.2). Recommended concentration: 0.3-1 ?g/mL.

RPGRIP1L antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RPGRIP1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR