Anti-LNX1 Goat Polyclonal Antibody

46-682
10117-024EA 494.34 USD
10117-024
Anti-LNX1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Preliminary experiments gave next to 70 kDa bands at approximately 75 kDa and 27 kDa in human kidney and lung lysates after 0.5 ?g/mL antibody staining. Please note that currently we cannot f

LNX1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LNX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR