Anti-KCNJ11 Goat Polyclonal Antibody

Supplier: Prosci
46-679
10112-202EA 701.94 USD
10112-202
Anti-KCNJ11 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 45-48 kDa band observed in human skeletal muscle lysates (calculated MW of 43.5 kDa according to NP_000516.3). Recommended concentration: 0.1-0.3 ?g/mL. Immunohistochemistry: In paraffin embedded human pancreas shows variable staining across the islet of Langerhans. Recommended concentration, 3-5 ?g/mL.

KCNJ11 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KCNJ11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR