Anti-GCGR Goat Polyclonal Antibody

46-667
10117-002EA 617.84 USD
10117-002
Anti-GCGR Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 35 kDa band in human kidney and liver lysates after 0.03 ?g/mL antibody staining. Please note that currently we cannot find an explanation in th

Glucagon Receptor antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GCGR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR