Anti-EPB41L4B Goat Polyclonal Antibody

Supplier: Prosci
46-654
10116-984EA 701.94 USD
10116-984
Anti-EPB41L4B Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 48 kDa band in human brain (hippocampus, Cerebellar cortex, cerebellum) and human bone marrow lysates after 0.3 ?g/mL antibody staining.

EPB41L4B antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: EPB41L4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR