Anti-CRHR1 Goat Polyclonal Antibody

Supplier: Prosci
46-648
10116-972EA 701.94 USD
10116-972
Anti-CRHR1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 60-70 kDa band observed in human cerebellum and human colon lysates (calculated MW of 47.7 kDa according to NP_004373.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Xu et al, Mol Pharmacol. 2001 Mar;59 (3) :485-92; PMID: 11179443). Recommended concentration: 0.1-0.3 ?g/mL. this antibody was successfully used in immunofluorescence on mouse brain at 2 ?g/mL when incubated at 4C for 40h.

CRF Receptor antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CRHR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR