Anti-CLIC4 Goat Polyclonal Antibody

Supplier: Prosci

46-645
10116-966EA 560.36 USD
10116-966
Anti-CLIC4 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64000. Western Blot: Approximately 30 kDa band observed in human kidney and human placenta lysates (calculated MW of 28.8 kDa according to NP_039234.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Edwards et al, Am J Physiol. 1999 Mar;276 (3 Pt 2) :F398-408.; PMID: 10070163). Recommended concentration: 0.1-0.3 ?g/mL.

CLIC4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CLIC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR