Anti-AVPR1A Goat Polyclonal Antibody

Supplier: Prosci
46-630
10116-940EA 701.94 USD
10116-940
Anti-AVPR1A Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 85 kDa band observed in human frontal cortex lysates and in rat brain lysates and in lysates of cell line NIH3T3 (calculated MW of 46.8 kDa according to NP_000697.1). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Phalipou et al, J Biol Chem. 1997 Oct 17;272(42):26536-44; PMID: 9334232). Recommended concentration: 0.03-0.1 ?g/mL.

AVPR1A antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AVPR1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR