Anti-ADRB3 Goat Polyclonal Antibody

Supplier: Prosci
46-623
10116-926EA 701.94 USD
10116-926
Anti-ADRB3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:128,000. Western Blot: Approximately 45 kDa band observed in human brain (cerebellum, cerebral cortex, frontal cortex) lysates (calculated MW of 43.5 kDa according to NP_000016.1). Recommended concentration: 0.03

ADRB3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ADRB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR