Anti-ZNF703 Goat Polyclonal Antibody

Supplier: Prosci
46-618
10116-916EA 701.94 USD
10116-916
Anti-ZNF703 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 28-30 kDa band in human breast lysates after 1 ?g/mL antibody staining. Approximately 60 kDa band observed in human breast carcinoma lysates (calculated MW of 58.2 kDa according to NP_079345.1). Recommended concentration: 1-3 ?g/mL.

ZNF703 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ZNF703
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR