Anti-ZNF217 Goat Polyclonal Antibody

Supplier: Prosci
46-616
10116-912EA 701.94 USD
10116-912
Anti-ZNF217 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16,000. Western Blot: Approximately 75 kDa band in human prostate, spleen, testis and thymus lysates after 0.03 ?g/mL antibody staining.1. The 75 kDa band was successfully blocked by incubation with the immunizing peptide.

ZNF217 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ZNF217
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR