Anti-CCDC50 Goat Polyclonal Antibody

Supplier: Prosci
46-607
10116-900EA 701.94 USD
10116-900
Anti-CCDC50 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 18 kDa band in human brain lysates after 1 ?g/mL antibody staining.3 kDa according to NP_848018.1 and 35.8 kDa according to NP_777568.1. The 18 kDa band was successfully blocked by incubation with the immunizing peptide. Immunohistochemistry: In paraffin embedded human pancreas shows strong staining in cytoplasm in select acinar cells. Recommended concentration, 5-10 ?g/mL.

Ymer antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CCDC50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR