Anti-WNT3 Goat Polyclonal Antibody

Supplier: Prosci
46-598
10116-888EA 701.94 USD
10116-888
Anti-WNT3 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:2000. Western Blot: Approximately 38 kDa band observed in human brain lysates (calculated MW of 39.6 kDa according to NP_110380.1). Recommended concentration: 1-3 ?g/mL. Immunohistochemistry: In paraffin embedded human breast shows strong fibrous staining in myoepithelial cells. Recommended concentration, 3-5 ?g/mL.

WNT3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: WNT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR