Anti-WIPF1 Goat Polyclonal Antibody

46-596
10116-884EA 689.45 USD
10116-884
Anti-WIPF1 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 55 kDa band observed in human lymph node, spleen and tonsil lysates (calculated MW of 51.3 kDa according to NP_003378.3 and NP_001070737.1). In transfected HEK293 transiently expressing WIPF1 a band of approximately 55 kDa is observed. This band is not observed in the non-transfected HEK293. Recommended concentration: 0.03-0.1 ?g/mL.

WIPF1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: WIPF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR