Anti-VPS35 Goat Polyclonal Antibody

Supplier: Prosci
46-584
10116-868EA 701.94 USD
10116-868
Anti-VPS35 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 90 kDa band observed in human brain and testis lysates (predicted MW of 91 kDa according to NP_060676.2). Recommended for use at 1-3 ?g/mL. Immunohistochemistry: In paraffin embedded human testis, shows positive staining in Leydig cells, and cytoplasmic staining (dot-like) of some epithelial cells of the epididymis and of the rete testis.

VPS35 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: VPS35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR