Anti-VPS25 Goat Polyclonal Antibody

Supplier: Prosci
46-578
10116-862EA 701.94 USD
10116-862
Anti-VPS25 Goat Polyclonal Antibody
Antibodies
ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Approximately 20-22 kDa kDa band observed in human duodenum, kidney and liver lysates (calculated MW of 20.7 kDa according to NP_115729.1). Recommended concentration: 0.5-1.5 ?g/mL.

VPS25 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: VPS25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR